Learn More
STEMCELL Technologies Human Recombinant PDGF-AA, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78095.1
Encompass_Preferred
Use platelet-derived growth factor (PDGF) AA to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into oligodendrocyte precursor cells (Piao et al. Cell Stem Cell, 2015). PDGF is a dimeric glycoprotein that signals through the receptor tyrosine kinases PDGFRα and PDGFRβ. It is a potent mitogen for cells of mesenchymal origin and induces migration through signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al. Growth Factors, 2007). PDGF-AA mediates proliferation of oligodendrocyte progenitor cells and oligodendrocyte lineage differentiation through the activation of extracellular signal-regulated kinases 1 and 2 (ERK1/2) (Hu et al. Neuroscience, 2008). Human recombinant PDGF-AA from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 50 ng/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: GDGF, Glioma-derived growth factor, ODGF, Osteosarcoma-derived growth factor, Platelet-derived growth factor-AA.
Catalog No. 50-197-6454
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.