Learn More
STEMCELL Technologies Human Recombinant R-Spondin-1, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78213.3
Encompass_Preferred
Use R-Spondin-1 (RSPO1) to modulate pleiotropic roles during embryogenesis, such as the specification of hematopoietic stem cells (Genthe & Clements. Development, 2017) and the growth, survival, and migration of ovarian cancer cells (Liu et al. Gene, 2019). RSPO1 is the prototype member of the R-Spondin (RSPO) protein subfamily of a superfamily of thrombospondin type 1 repeat (TSR-1)-containing proteins (Kim et al. Cell Cycle, 2006). Although unable to initialize signaling, RSPO family members are potent enhancers of WNT signaling (Cruciat & Niehrs. Cold Spring Harb Perspect Biol, 2013). Through WNT signaling, RSPO1 activates β­-catenin signaling and indirectly increases low-density lipoprotein receptor-related protein on the cell surface (Binnerts et al. Proc Natl Acad Sci USA, 2007). Human recombinant R-Spondin-1 from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 50 ng/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: hRspo1, Roof plate-specific Spondin-1.
Catalog No. 50-197-6751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.