Learn More
STEMCELL Technologies Human Recombinant TNF-alpha, Size: 50 ug
SDP
Supplier:  STEMCELL Technologies 78068
Encompass_Preferred
Tumor necrosis factor-a (TNF-a) is a pro-inflammatory cytokine that activates NF-kB MAPK and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-a in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-a among them B cells NK cells mast cells neutrophils dendritic cells microglia endothelial cells smooth muscle cells cardiomyocytes and fibroblasts. TNF-a has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immuno- suppressive responses. TNF-a stimulates expression of E- and P-selectins thus facilitating adhesion of neutrophils monocytes and memory T cells to activated platelets and endothelial cells (Zelová Hosek). Other effects of TNF-a include vasodilatation and edema formation.
Catalog No. 50-239-0179
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.