Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Human Regulatory T Cell Staining Kit

Catalog No. 501128873
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-8873 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-8873 Supplier Invitrogen™ Supplier No. 88899940
  • $748.00
    $560.65 / Each of 1

    Save $187.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide and formaldehyde

Includes

  • Flow Cytometry Staining Buffer
  • Fixation/Permeabilization Concentrate (4X)
  • Fixation/Permeabilization Diluent
  • Permeabilization Buffer (10X)
  • Normal Rat Serum
  • eZFluor™ Anti-Human CD4 FITC and CD25 APC Cocktail (clones RPA-T4 and BC96, cat. 22-0425): 25 tests Light-sensitive material.
  • Anti-Human Foxp3 PE (clone PCH101, cat. 12-4776): 25 tests Light-sensitive material.
  • Rat IgG2a K Isotype Control PE : 25 tests Light-sensitive material.

Contains all of the buffers and monoclonal antibodies for CD4, CD25, and Foxp3 necessary to successfully stain and identify regulatory T cells from human peripheral blood cells.

Tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. The PCH101 antibody and the Rat IgG2a K Isotype Control can be used at 20μL (0.25μg) per test.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
Quantity 1 kit
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Cell Staining Kit
I want to obtain good results, so which format of Ebioscience Foxp3 antibody should I order?

Our recommendation is to use the Foxp3 antibodies in PE or APC as they are brighter than FITC. Furthermore we prefer CD4 in FITC since the expression levels are high enough to use with this fluorochrome. We then use CD25 in APC. Please see our Treg staining Kits for prebundled kits with optimal fluorochrome combinations (Cat. No. 88-8999-40 and 88-8111-40).

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.