Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
The Human Relaxin-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Relaxin-2.
Specifications
Specifications
| Assay Sensitivity | 0pg/mL |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Kit Contents | Capture antibody, Detector antibody, Recombinant standard, Streptavidin-HRP |
| Quantity | 5 Plate |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
