Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Relaxin-2 Recombinant Protein, PeproTech®

Catalog No. 130155UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-15-5UG 5 μg
13015100UG 100 μg
130-15-1MG 1 mg
130-15-25UG 25 μg
13015500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 130-15-5UG Supplier Gibco™ Supplier No. 130155UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Relaxin-2 is a non-glycosylated, 6.0 kDa, disulfide-linked, heterodimeric protein consisting of a 24 amino acid A-chain, and a 29 amino acid B-chain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Relaxin 1 (also referred to as Relaxin or RLXH1) is a peptide hormone produced by the corpora lutea of the ovary during pregnancy in many mammalian species, including human. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, Relaxin 1 may influence the timing of parturition. Like insulin, Relaxin 1 consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the amino-terminus. The gene that encodes the human Relaxin 1 protein maps to chromosome 9. Relaxin 2, a related protein, is selectively expressed in the ovary during pregnancy. The gene that encodes the human Relaxin 2 protein also maps to chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04090
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6019
Molecular Weight (g/mol) 6 kDa
Name Human Relaxin-2
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias bA12D24.1.1; bA12D24.1.2; H2; H2-preprorelaxin; H2-RLX; Prorelaxin H2; relaxin 2; Relaxin A chain; Relaxin B chain; relaxin H2; relaxin, ovarian, of pregnancy; RLN2; RLX H2-RLX; RLXH2
Common Name Relaxin 2
Gene Symbol RLN2
Conjugate Unconjugated
Recombinant Recombinant
Sequence A-Chain: QLYSALANKC CHVGCTKRSL ARFC B-Chain: DSWMEEVIKL CGRELVRAQI AICGMSTWS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.