Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Renin Quantikine ELISA Kit
SDP

Catalog No. DREN00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DREN00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DREN00 Supplier R&D Systems™ Supplier No. DREN00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This assay detects pro- and mature Renin.

The Human Renin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Renin in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), urine (50 ul).

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 14.8 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human Renin. This assay recognizes both the Pro- and mature versions of Recombinant Human Renin.
For Use With (Application) ELISA
Gene ID (Entrez) 5972
Gene Symbol REN
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, REN, renin, renin precursor, renal
Target Renin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.