Learn More
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Gene ID (Entrez) | 56729 |
| Product Line | R&D Systems |
| Synonym | Adipose tissue-specific secretory factor, ADSF, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN, RETN1, RSTNXCP1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
