Learn More
Walter Products™ Human Respiratory System Model with Magnified Alveolus
SCI_ED
Catalog No. S139156 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
21 x 15 x 2.5 in.
Catalog No. Dimensions (L x W x H)
S139156 21 x 15 x 2.5 in.
1 options
This item is not returnable. View return policy
Catalog No. S139156 Supplier Walter Products™ Supplier No. B10446
Add to Cart
Add to Cart
This item is not returnable. View return policy
Deluxe model of human respiratory system featuring key anatomical features labeled with accompanying key card.
  • Relief model shows the human respiratory system as well as a 150 times magnified alveolus with blood vessels
  • Regions shown include the oronasal cavity, larynx and trachea, primary bronchi and bronchial tree
  • Upper respiratory section is medially sectioned to show the interior anatomical details
  • Both are mounted on a base
Specifications
Dimensions (L x W x H) 21 x 15 x 2.5 in.
Material PVC
Product Type Anatomical Model