Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RGC32 Partial ORF (NP_054778.1, 1 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959868 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-868 25 μg
89-959-867 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-868 Supplier Abnova™ Supplier No. H00028984Q02L

Used for AP, Array, ELISA, WB-Re

This gene is thought to regulate cell cycle progression. It is induced by p53 in response to DNA damage, or by sublytic levels of complement system proteins that result in activation of the cell cycle. The encoded protein localizes to the cytoplasm during interphase and to centrosomes during mitosis. The protein forms a complex with polo-like kinase 1. The protein also translocates to the nucleus in response to treatment with complement system proteins, and can associate with and increase the kinase activity of cell division cycle 2 protein. In different assays and cell types, overexpression of this protein has been shown to activate or suppress cell cycle progression. [provided by RefSeq]

Sequence: MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM

Specifications

Accession Number NP_054778.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 28984
Molecular Weight (g/mol) 38.61kDa
Name RGC32 (Human) Recombinant Protein (Q02)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0564/MGC87338/RGC-32/RGC32/bA157L14.2
Common Name C13orf15
Gene Symbol C13orf15
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.