Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RGM-C/Hemojuvelin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7158780
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3720SP 25 μg
MAB3720100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3720SP Supplier R&D Systems Supplier No. MAB3720SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RGM-C/Hemojuvelin Monoclonal specifically detects RGM-C/Hemojuvelin in Human samples. It is validated for ELISA.

Specifications

Antigen RGM-C/Hemojuvelin
Applications ELISA
Classification Monoclonal
Clone 751741
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q8ZVN8
Gene Alias DL-M, haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2, HFE2AMGC23953, HJV, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMC, RGMCRGM domain family member C
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human RGM-C/Hemojuvelin, Gln36-Asp400, Accession No. Q8ZVN8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 148738.0
Test Specificity Detects human RGM-C/Hemojuvelin in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.