Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RGP1 Synthetic Peptide

Catalog No. PEP0639
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0639 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0639 Supplier Invitrogen™ Supplier No. PEP0639
Add to Cart
Add to Cart

Synthetic Peptide

RGP1 Synthetic Peptide, PEP-0639, from Invitrogen.

Retrograde golgi transport homolog 1 (RGP1) is the mammalian homolog to the yeast RGP1, a protein that forms a tight complex with RIC1. This complex binds Ypt6p and stimulates guanine nucleotide exchange. RGP1 is localized to the Golgi and is thought to be a potential Golgi recycling factor. Rgp1 yeast mutants exhibit defects in retrograde trafficking similar to those seen in yeast with mutations in other retrograde Golgi transport proteins. It is expected that RGP1 plays a similar role in mammalian cells to that seen in yeast.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9827
Species Human
Purification Method purified
Accession Number Q92546
Common Name Human RGP1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol RGP1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.