Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RGS11 Partial ORF (NP_003825, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-890 25 μg
89-959-889 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-890 Supplier Abnova™ Supplier No. H00008786Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Sequence: DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMT

Specifications

Accession Number NP_003825
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8786
Molecular Weight (g/mol) 36.63kDa
Name RGS11 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias RS11
Common Name RGS11
Gene Symbol RGS11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.