Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RGS13 Full-length ORF (AAH16667, 1 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-900 10 ug
89-959-901 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-900 Supplier Abnova™ Supplier No. H00006003P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the regulator of G protein signaling (RGS) family. RGS family members share similarity with S. cerevisiae SST2 and C. elegans egl-10 proteins, which contain a characteristic conserved RGS domain. RGS proteins accelerate GTPase activity of G protein alpha-subunits, thereby driving G protein into their inactive GDP-bound form, thus negatively regulating G protein signaling. RGS proteins have been implicated in the fine tuning of a variety of cellular events in response to G protein-coupled receptor activation. The biological function of this gene, however, is unknown. Two transcript variants encoding the same isoform exist. [provided by RefSeq]

Sequence: MSRRNCWICKMCRDESKRPPSNLTLEEVLQWAQSFENLMATKYGPVVYAAYLKMEHSDENIQFWMACETYKKIASRWSRISRAKKLYKIYIQPQSPREINIDSSTRETIIRNIQEPTETCFEEAQKIVYMHMERDSYPRFLKSEMYQKLLKTMQSNNSF

Specifications

Accession Number AAH16667
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6003
Molecular Weight (g/mol) 43.23kDa
Name RGS13 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSRRNCWICKMCRDESKRPPSNLTLEEVLQWAQSFENLMATKYGPVVYAAYLKMEHSDENIQFWMACETYKKIASRWSRISRAKKLYKIYIQPQSPREINIDSSTRETIIRNIQEPTETCFEEAQKIVYMHMERDSYPRFLKSEMYQKLLKTMQSNNSF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC17173
Common Name RGS13
Gene Symbol RGS13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.