Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RGS17 Full-length ORF (AAH13117, 1 a.a. ∽210 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-914 10 μg
89-959-915 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-914 Supplier Abnova™ Supplier No. H00026575P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. [provided by RefSeq]

Sequence: MRKRQQSQNEGTPAVSQAPGNQRPNNTCCFCWCCCCSCSCLTVRNEERGENAGRPTHTTKMESIQVLEECQNPTAEEVLSWSQNFDKMMKAPAGRNLFREFLRTEYSEENLLFWLACEDLKKEQNKKVIEEKARMIYEDYISILSPKEVSLDSRVREVINRNLLDPNPHMYEDAQLQIYTLMHRDSFPRFLNSQIYKSFVESTAGSSSES

Specifications

Accession Number AAH13117
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26575
Molecular Weight (g/mol) 48.73kDa
Name RGS17 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MRKRQQSQNEGTPAVSQAPGNQRPNNTCCFCWCCCCSCSCLTVRNEERGENAGRPTHTTKMESIQVLEECQNPTAEEVLSWSQNFDKMMKAPAGRNLFREFLRTEYSEENLLFWLACEDLKKEQNKKVIEEKARMIYEDYISILSPKEVSLDSRVREVINRNLLDPNPHMYEDAQLQIYTLMHRDSFPRFLNSQIYKSFVESTAGSSSES
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias RGS-17/RGSZ2/hRGS17
Common Name RGS17
Gene Symbol RGS17
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.