Learn More
ATCC Human Rhinovirus 6 (ATCC VR-486) - Frozen Research Strain for Respiratory and Infectious Disease Studies
Supplier: ATCC VR486
Human rhinovirus 6 (VR-486) is a frozen research strain supplied for infectious disease and respiratory research. The isolate (strain Thompson) has been whole-genome sequenced, is typically propagated in HeLa cells, produces a cytopathic effect, and requires storage at -70°C or colder.
- Classification: Picornaviridae, rhinovirus, human rhinovirus B.
- Strain designation: Thompson.
- Product format: Frozen; store at -70°C or colder.
- Genome sequenced: Yes.
- Isolation source: Human throat washings.
- Propagation host: HeLa cells (CCL-2).
- Host range: Human cell lines; some MkK cells.
- Effects: Cytopathic effect (CPE).
- Incubation: 1-7 days.
- Mycoplasma: Not detected.
- Quality control: Whole-genome sequencing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.