Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RHOG Full-length ORF (NP_001656.2, 1 a.a. - 191 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-993 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-959-993 Supplier Abnova™ Supplier No. H00000391P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

ARHG is a member of the RAS superfamily of genes, which encode GTP-binding proteins that act in the pathway of signal transduction and play a key role in the regulation of cellular functions.[supplied by OMIM]

Sequence: MQSIKCVVVGDGAVGKTCLLICYTTNAFPKEYIPTVFDNYSAQSAVDGRTVNLNLWDTAGQEEYDRLRTLSYPQTNVFVICFSIASPPSYENVRHKWHPEVCHHCPDVPILLVGTKKDLRAQPDTLRRLKEQGQAPITPQQGQALAKQIHAVRYLECSALQQDGVKEVFAEAVRAVLNPTPIKRGRSCILL

Specifications

Accession Number NP_001656.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 391
Molecular Weight (g/mol) 47.7kDa
Name RHOG (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MQSIKCVVVGDGAVGKTCLLICYTTNAFPKEYIPTVFDNYSAQSAVDGRTVNLNLWDTAGQEEYDRLRTLSYPQTNVFVICFSIASPPSYENVRHKWHPEVCHHCPDVPILLVGTKKDLRAQPDTLRRLKEQGQAPITPQQGQALAKQIHAVRYLECSALQQDGVKEVFAEAVRAVLNPTPIKRGRSCILL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ARHG/MGC125835/MGC125836
Common Name RHOG
Gene Symbol RHOG
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.