Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RIPK2/RIP2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10003295
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10003295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10003295 Supplier R&D Systems Supplier No. FAB103872V100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

RIPK2 Monoclonal antibody specifically detects RIPK2 in Human samples. It is validated for Western Blot

Specifications

Antigen RIPK2
Applications Western Blot
Classification Monoclonal
Clone 2527A
Conjugate Alexa Fluor 405
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias CARD3, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARDIAKCCK, EC 2.7.11, EC 2.7.11.1, GIG30, growth-inhibiting gene 30, receptor interacting protein 2, receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase, Receptor-interacting protein 2, receptor-interacting serine/threonine-protein kinase 2, receptor-interacting serine-threonine kinase 2, RICK, RICKGIG30, RIP2, RIP-2, RIP2CARD-carrying kinase, RIP-like-interacting CLARP kinase
Host Species Rabbit
Immunogen E.coli-derived recombinant human RIPK2/RIP2, Met1-Met540, Accession # O43353
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8767
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Product Type Antibody
Form Purified
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.