Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RIPK3/RIP3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB76042SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB76042SP 25 μg
MAB76042100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB76042SP Supplier R&D Systems Supplier No. MAB76042SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RIPK3/RIP3 Monoclonal specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen RIPK3/RIP3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1312D
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Immunocytochemistry 1-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9Y572
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Host Species Rabbit
Immunogen Synthetic peptide containing human RIPK3/RIP3, Accession # Q9Y572
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 11035
Test Specificity Detects human RIPK3/RIP3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.