Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RNF12 Partial ORF (NP_057204, 1 a.a. - 83 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-162 10 μg
89-960-163 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-960-162 Supplier Abnova™ Supplier No. H00051132Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a RING-H2 zinc finger protein. It has been shown to be an E3 ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDEN

Specifications

Accession Number NP_057204
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51132
Molecular Weight (g/mol) 34.87kDa
Name RNF12 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDEN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC15161/NY-REN-43/RLIM
Common Name RNF12
Gene Symbol RNF12
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.