Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RNF36 Full-length ORF (NP_542783.2, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-328 25 μg
89-960-327 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-960-328 Supplier Abnova™ Supplier No. H00140691P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: MEEELAIQQGQLETTLKELQTLRNMQKEAIAAHKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEQGMKVLATRELISRKLNLGQYKGPIQYMVWREMQDTLCPGLSPLTLDPKTAHPNLVLSKSQTSVWHGDIKKIMPDDPERFDSSVAVLGSRGFTSGKWYWEVEVAKKTKWTVGVVRESIIRKGSCPLTPEQGFWLLRLRNQTDLKALDLPSFSLTLTNNLDKVGIYLDYEGGQLSFYNAKTMTHIYTFSNTFMEKLYPYFCPCLNDGGENKEPLHILHPQ

Specifications

Accession Number NP_542783.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 140691
Molecular Weight (g/mol) 65.7kDa
Name RNF36 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MEEELAIQQGQLETTLKELQTLRNMQKEAIAAHKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEQGMKVLATRELISRKLNLGQYKGPIQYMVWREMQDTLCPGLSPLTLDPKTAHPNLVLSKSQTSVWHGDIKKIMPDDPERFDSSVAVLGSRGFTSGKWYWEVEVAKKTKWTVGVVRESIIRKGSCPLTPEQGFWLLRLRNQTDLKALDLPSFSLTLTNNLDKVGIYLDYEGGQLSFYNAKTMTHIYTFSNTFMEKLYPYFCPCLNDGGENKEPLHILHPQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HSD34/RNF36/Trif
Common Name TRIM69
Gene Symbol TRIM69
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.