Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RNF40 Partial ORF (NP_055586, 102 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-339 10 μg
89-960-340 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-960-339 Supplier Abnova™ Supplier No. H00009810Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery. [provided by RefSeq]

Sequence: DETVEALLRCHESQGELSSAPEAPGTQEGPTCDGTPLPEPGTSELRDPLLMQLRPPLSEPALAFVVALGASSSEEVELELQGRMEFSKAAVSRVVEASD

Specifications

Accession Number NP_055586
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9810
Molecular Weight (g/mol) 36.63kDa
Name RNF40 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen DETVEALLRCHESQGELSSAPEAPGTQEGPTCDGTPLPEPGTSELRDPLLMQLRPPLSEPALAFVVALGASSSEEVELELQGRMEFSKAAVSRVVEASD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BRE1B/DKFZp686K191/KIAA0661/MGC13051/RBP95/STARING
Common Name RNF40
Gene Symbol RNF40
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.