Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RNPC3 Full-length ORF (AAH10697, 1 a.a. - 261 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-364 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-960-364 Supplier Abnova™ Supplier No. H00055599P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Two types of spliceosomes catalyze splicing of pre-mRNAs. The major U2-type spliceosome is found in all eukaryotes and removes U2-type introns, which represent more than 99% of pre-mRNA introns. The minor U12-type spliceosome is found in some eukaryotes and removes U12-type introns, which are rare and have distinct splice consensus signals. The U12-type spliceosome consists of several small nuclear RNAs and associated proteins. This gene encodes a 65K protein that is a component of the U12-type spliceosome. This protein contains two RNA recognition motifs (RRMs), suggesting that it may contact one of the small nuclear RNAs of the minor spliceosome. [provided by RefSeq]

Sequence: MNKLMELANLQPKRPKTIKQRHVRKKRKIKDMLNTPLCPSHSSLHPVLLPSDVFDQPQPVGNKRIEFHISTDMPAAFKKDLEKEQNCEEKNHDLPATEVDASNIGFGKIFPKPNLDITEEIKEDSDEMPSECISRRELEKGRISREEMETLSVFRSYEPGEPNCRIYVKNLAKHVQEKDLKYIFGRYVDFSSETQRIMFDIRLMKEGRMKGQAFIGLPNEKAAAKALKEANGYVLFGKPMVVQFARSARPKQDPKEGKRKC

Specifications

Accession Number AAH10697
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55599
Molecular Weight (g/mol) 54.45kDa
Name RNPC3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MNKLMELANLQPKRPKTIKQRHVRKKRKIKDMLNTPLCPSHSSLHPVLLPSDVFDQPQPVGNKRIEFHISTDMPAAFKKDLEKEQNCEEKNHDLPATEVDASNIGFGKIFPKPNLDITEEIKEDSDEMPSECISRRELEKGRISREEMETLSVFRSYEPGEPNCRIYVKNLAKHVQEKDLKYIFGRYVDFSSETQRIMFDIRLMKEGRMKGQAFIGLPNEKAAAKALKEANGYVLFGKPMVVQFARSARPKQDPKEGKRKC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ20008/FLJ25070/KIAA183/RBM40/RNP
Common Name RNPC3
Gene Symbol RNPC3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.