Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ROD1 Full-length ORF (AAH44585.1, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960395 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-395 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-960-395 Supplier Abnova™ Supplier No. H00009991P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

ROD1 is a functional homolog of nrd1, an S. pombe RNA-binding protein that suppresses the onset of differentiation.[supplied by OMIM]

Sequence: MDASPSPFSLPKKLNELSARRGSDELLSSGIINGPFTMNSSTPSTANGNDSKKFKRDRPPCSPSRVLHLRKIPCDVTEAEIISLGLPFGKVTNLLMLKGKSQAFLEMASEEAAVTMVNYYTPITPHLRSQPVYIQYSNHRELKTDNLPNQARAQAALQAVSAVQSGSLALSGGPSNEGTVLPGQSPVLRIIIENLFYPVTLEVLHQIFSKFGTVLKIITFTKNNQFQALLQYADPVNAHYAKMALDGQNIYNACCTLRIDFSKLTSLNVKYNNDKSRDFTRLDLPTGDGQPSLEPPMAAAFGAPGIISSPYAGAAGFAPAIGFPQATGLSVPAVPGALGPLTITSSAVTGRMAIPGASGIPGNSVLLVTNLNPDLITPHGLFILFGVYGDVHRVKIMFNKKENALVQMADANQAQLAMNHLSGQRLYGKVLRATLSKHQAVQLPREGQEDQGLTKDFSNSPLHRFKKPGSKNFQNIFPPSATLHLSNIPPSVTVDDLKNLFIEAGCSVKAFKFFQKDRKMALIQLGSVEEAIQALIELHNHDLGENHHLRVSFSKSTI

Specifications

Accession Number AAH44585.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9991
Molecular Weight (g/mol) 86.8kDa
Name ROD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MDASPSPFSLPKKLNELSARRGSDELLSSGIINGPFTMNSSTPSTANGNDSKKFKRDRPPCSPSRVLHLRKIPCDVTEAEIISLGLPFGKVTNLLMLKGKSQAFLEMASEEAAVTMVNYYTPITPHLRSQPVYIQYSNHRELKTDNLPNQARAQAALQAVSAVQSGSLALSGGPSNEGTVLPGQSPVLRIIIENLFYPVTLEVLHQIFSKFGTVLKIITFTKNNQFQALLQYADPVNAHYAKMALDGQNIYNACCTLRIDFSKLTSLNVKYNNDKSRDFTRLDLPTGDGQPSLEPPMAAAFGAPGIISSPYAGAAGFAPAIGFPQATGLSVPAVPGALGPLTITSSAVTGRMAIPGASGIPGNSVLLVTNLNPDLITPHGLFILFGVYGDVHRVKIMFNKKENALVQMADANQAQLAMNHLSGQRLYGKVLRATLSKHQAVQLPREGQEDQGLTKDFSNSPLHRFKKPGSKNFQNIFPPSATLHLSNIPPSVTVDDLKNLFIEAGCSVKAFKFFQKDRKMALIQLGSVEEAIQALIELHNHDLGENHHLRVSFSKSTI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781I1117/PTBP3
Common Name ROD1
Gene Symbol ROD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.