Learn More
Sino Biological Human RPS6 PAb 100UL 110595-T32-100
Supplier:  Sino Biological 110595T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to residues around (Ser235, 236) of Human Phospho-S6 Ribosomal Protein, and purified by antigen affinity chromatography.
Catalog No. 50-241-0103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.