Learn More
Raybiotech Inc HUMAN RTK PHOSPHO-ARRAY C1 (8)
Supplier:  Raybiotech Inc AAHPRTK18
Encompass_Preferred
RayBio C-Series Human Receptor Tyrosine Kinase Phosphorylation Antibody Array 1 Kit. Detects 71 Human RTKs. Suitable for all liquid sample types but intended for use with cell and tissue lysates.
Catalog No. 50-104-3167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.