Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RTN4 Full-length ORF (NP_065393.1, 1 a.a. - 1192 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-896 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-960-896 Supplier Abnova™ Supplier No. H00057142P02
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. The product of this gene is a potent neurite outgrowth inhibitor which may also help block the regeneration of the central nervous system in higher vertebrates. Alternatively spliced transcript variants derived both from differential splicing and differential promoter usage and encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MEDLDQSPLVSSSDSPPRPQPAFKYQFVREPEDEEEEEEEEEEDEDEDLEELEVLERKPAAGLSAAPVPTAPAAGAPLMDFGNDFVPPAPRGPLPAAPPVAPERQPSWDPSPVSSTVPAPSPLSAAAVSPSKLPEDDEPPARPPPPPPASVSPQAEPVWTPPAPAPAAPPSTPAAPKRRGSSGSVDETLFALPAASEPVIRSSAENMDLKEQPGNTISAGQEDFPSVLLETAASLPSLSPLSAASFKEHEYLGNLSTVLPTEGTLQENVSEASKEVSEKAKTLLIDRDLTEFSELEYSEMGSSFSVSPKAESAVIVANPREEIIVKNKDEEEKLVSNNILHNQQELPTALTKLVKEDEVVSSEKAKDSFNEKRVAVEAPMREEYADFKPFERVWEVKDSKEDSDMLAAGGKIESNLESKVDKKCFADSLEQTNHEKDSESSNDDTSFPSTPEGIKDRSGAYITCAPFNPAATESIATNIFPLLGDPTSENKTDEKKIEEKKAQIVTEKNTSTKTSNPFLVAAQDSETDYVTTDNLTKVTEEVVANMPEGLTPDLVQEACESELNEVTGTKIAYETKMDLVQTSEVMQESLYPAAQLCPSFEESEATPSPVLPDIVMEAPLNSAVPSAGASVIQPSSSPLEASSVNYESIKHEPENPPPYEEAMSVSLKKVSGIKEEIKEPENINAALQETEAPYISIACDLIKETKLSAEPAPDFSDYSEMAKVEQPVPDHSELVEDSSPDSEPVDLFSDDSIPDVPQKQDETVMLVKESLTETSFESMIEYENKEKLSALPPEGGKPYLESFKLSLDNTKDTLLPDEVSTLSKKEKIPLQMEELSTAVYSNDDLFISKEAQIRETETFSDSSPIEIIDEFPTLISSKTDSFSKLAREYTDLEVSHKSEIANAPDGAGSLPCTELPHDLSLKNIQPKVEEKISFSDDFSKNGSATSKVLLLPPDVSALATQAEIESIVKPKVLVKEAEKKLPSDTEKEDRSPSAIFSAELSKTSVVDLLYWRDIKKTGVVFGASLFLLLSLTVFSIVSVTAYIALALLSVTISFRIYKGVIQAIQKSDEGHPFRAYLESEVAISEELVQKYSNSALGHVNCTIKELRRLFLVDDLVDSLKFAVLMWVFTYVGALFNGLTLLILALISLFSVPVIYERHQAQIDHYLGLANKNVKDAMAKIQAKIPGLKRKAE

Specifications

Accession Number NP_065393.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57142
Molecular Weight (g/mol) 156.3kDa
Name RTN4 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MEDLDQSPLVSSSDSPPRPQPAFKYQFVREPEDEEEEEEEEEEDEDEDLEELEVLERKPAAGLSAAPVPTAPAAGAPLMDFGNDFVPPAPRGPLPAAPPVAPERQPSWDPSPVSSTVPAPSPLSAAAVSPSKLPEDDEPPARPPPPPPASVSPQAEPVWTPPAPAPAAPPSTPAAPKRRGSSGSVDETLFALPAASEPVIRSSAENMDLKEQPGNTISAGQEDFPSVLLETAASLPSLSPLSAASFKEHEYLGNLSTVLPTEGTLQENVSEASKEVSEKAKTLLIDRDLTEFSELEYSEMGSSFSVSPKAESAVIVANPREEIIVKNKDEEEKLVSNNILHNQQELPTALTKLVKEDEVVSSEKAKDSFNEKRVAVEAPMREEYADFKPFERVWEVKDSKEDSDMLAAGGKIESNLESKVDKKCFADSLEQTNHEKDSESSNDDTSFPSTPEGIKDRSGAYITCAPFNPAATESIATNIFPLLGDPTSENKTDEKKIEEKKAQIVTEKNTSTKTSNPFLVAAQDSETDYVTTDNLTKVTEEVVANMPEGLTPDLVQEACESELNEVTGTKIAYETKMDLVQTSEVMQESLYPAAQLCPSFEESEATPSPVLPDIVMEAPLNSAVPSAGASVIQPSSSPLEASSVNYESIKHEPENPPPYEEAMSVSLKKVSGIKEEIKEPENINAALQETEAPYISIACDLIKETKLSAEPAPDFSDYSEMAKVEQPVPDHSELVEDSSPDSEPVDLFSDDSIPDVPQKQDETVMLVKESLTETSFESMIEYENKEKLSALPPEGGKPYLESFKLSLDNTKDTLLPDEVSTLSKKEKIPLQMEELSTAVYSNDDLFISKEAQIRETETFSDSSPIEIIDEFPTLISSKTDSFSKLAREYTDLEVSHKSEIANAPDGAGSLPCTELPHDLSLKNIQPKVEEKISFSDDFSKNGSATSKVLLLPPDVSALATQAEIESIVKPKVLVKEAEKKLPSDTEKEDRSPSAIFSAELSKTSVVDLLYWRDIKKTGVVFGASLFLLLSLTVFSIVSVTAYIALALLSVTISFRIYKGVIQAIQKSDEGHPFRAYLESEVAISEELVQKYSNSALGHVNCTIKELRRLFLVDDLVDSLKFAVLMWVFTYVGALFNGLTLLILALISLFSVPVIYERHQAQIDHYLGLANKNVKDAMAKIQAKIPGLKRKAE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ASY/NI220/250/NOGO/NOGO-A/NOGOC/NSP/NSP-CL/Nbla00271/Nbla10545/Nogo-B/Nogo-C/RTN-X/RTN4-A/RTN4-B1/RTN4-B2/RTN4-C
Common Name RTN4
Gene Symbol RTN4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.