Learn More
Invitrogen™ Human S100 P ELISA Kit
Description
Human S100 P quantitates human S100 P in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human S100 P.
Specifications
Specifications
| Accession Number | MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100P |
| Assay Range | 14 to 3,500 pg/mL |
| Assay Sensitivity | 14 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6286 |
| Gene Symbol | S100P |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.