Learn More
ACROBiosystems ACROBiosystems Human S100A10 Protein, His Tag
Supplier:  ACROBiosystems S10H5127100UG
Encompass_Preferred
Recombinant Protein;100UG;Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession # AAH15973).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.0 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-6752
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.