Learn More
R&D Systems™ Human S100A8/S100A9 Heterodimer Quantikine ELISA Kit

Shop All Novus Biologicals Products


Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Assay Range | 0.63 - 40ng/mL |
| Assay Sensitivity | 0.215ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested. |
| Sample Type | Cell Culture Supernatant, Fecal Extract, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Tissue Lysates, Urine |
| Specificity | Natural and Recombinant Human S100A8/S100A9 Heterodimer. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (10uL),Urine (50uL),Fecal Extract (50uL),Human Milk (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.