Learn More
Invitrogen™ Human S100B Recombinant Protein
Description
RP-75721 contains purified recombinant human S100 beta Chain protein with an N-terminal hexahistidine tag. RP-75721 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P04271 |
| Concentration | 0.1 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | PBS with 50% glycerol and no preservative; pH 7.4 |
| Gene ID (Entrez) | 6285 |
| Molecular Weight (g/mol) | 14.7 kDa |
| Name | Human S100B |
| pH Range | 7.4 |
| Quantity | 10 μg |
| Source | E. coli |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.