Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SAA1 (P02735, 122 amino acids) Partial Recombinant Protein

Catalog No. 89961008 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-008 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-008 Supplier Abnova™ Supplier No. P4022

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE

Specifications

Accession Number P02735
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 6288
Molecular Weight (g/mol) 11.5kDa
Name SAA1 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing SDS-PAGE Result
Quantity 50 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 0.1% acetic acid, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias MGC111216/PIG4/SAA/TP53I4
Common Name SAA1
Gene Symbol SAA1
Biological Activity For most in vitro applications, apo-SAA exerts its biological activity in the concentration range of 10-50μg/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.