Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SC5DL Full-length ORF (NP_001020127.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961113 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-113 25 ug
89-961-112 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-113 Supplier Abnova™ Supplier No. H00006309P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an enzyme of cholesterol biosynthesis. The encoded protein catalyzes the conversion of lathosterol into 7-dehydrocholesterol. Mutations in this gene have been associated with lathosterolosis. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq]

Sequence: MDLVLRVADYYFFTPYVYPATWPEDDIFRQAISLLIVTNVGAYILYFFCATLSYYFVFDHALMKHPQFLKNQVRREIKFTVQALPWISILTVALFLLEIRGYSKLHDDLGEFPYGLFELVVSIISFLFFTDMFIYWIHRGLHHRLVYKRLHKPHHIWKIPTPFASHAFHPIDGFLQSLPYHIYPFIFPLHKVVYLSLYILVNIWTISIHDGDFRVPQILQPFINGSAHHTDHHMFFDYNYGQYFTLWDRIGGSFKNPSSFEGKGPLSYVKEMTEGKRSSHSGNGCKNEKLFNGEFTKTE

Specifications

Accession Number NP_001020127.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6309
Molecular Weight (g/mol) 61.7kDa
Name SC5DL (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDLVLRVADYYFFTPYVYPATWPEDDIFRQAISLLIVTNVGAYILYFFCATLSYYFVFDHALMKHPQFLKNQVRREIKFTVQALPWISILTVALFLLEIRGYSKLHDDLGEFPYGLFELVVSIISFLFFTDMFIYWIHRGLHHRLVYKRLHKPHHIWKIPTPFASHAFHPIDGFLQSLPYHIYPFIFPLHKVVYLSLYILVNIWTISIHDGDFRVPQILQPFINGSAHHTDHHMFFDYNYGQYFTLWDRIGGSFKNPSSFEGKGPLSYVKEMTEGKRSSHSGNGCKNEKLFNGEFTKTE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ERG3/S5DES/SC5D
Common Name SC5DL
Gene Symbol SC5DL
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.