Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SCAMP4 Synthetic Peptide

Catalog No. PEP0901
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0901 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0901 Supplier Invitrogen™ Supplier No. PEP0901
Add to Cart
Add to Cart

Synthetic Peptide

SCAMP4 Synthetic Peptide, PEP-0901, from Invitrogen.

SCAMPs, or secretory carrier membrane proteins, are membrane proteins that are widely distributed and play a role in membrane trafficking. Most SCAMPs, such as SCAMP1, have N-terminal cytoplasmic NPF repeats, 4 central transmembrane regions, and a short C-terminal cytoplasmic tail. These NPF repeats are likely involved in mediating endocytosis. However, other SCAMPs, like SCAMP4, lack the NPF repeats and are thus unlikely to function in endocytosis. SCAMP4 has been associated with diseases such as Neurodevelopmental Disorder With Brain Abnormalities, Poor Growth, And Dysmorphic Facies and Strabismus.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 113178
Species Human
Purification Method purified
Accession Number Q969E2
Common Name Human SCAMP4
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol SCAMP4
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.