Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SCARB2 Full-length ORF (NP_005497.1, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-137 10 ug
89-961-138 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-137 Supplier Abnova™ Supplier No. H00000950P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a type III glycoprotein that is located primarily in limiting membranes of lysosomes and endosomes. Studies of the similar protein in mice and rat suggested that this protein may participate in membrane transportation and the reorganization of endosomal/lysosomal compartment. Deficiency of the similar protein in mice was reported to impair cell membrane transport processes and cause pelvic junction obstruction, deafness, and peripheral neuropathy. [provided by RefSeq]

Sequence: MGRCCFYTAGTLSLLLLVTSVTLLVARVFQKAVDQSIEKKIVLRNGTEAFDSWEKPPLPVYTQFYFFNVTNPEEILRGETPRVEEVGPYTYRELRNKANIQFGDNGTTISAVSNKAYVFERDQSVGDPKIDLIRTLNIPVLTVIEWSQVHFLREIIEAMLKAYQQKLFVTHTVDELLWGYKDEILSLIHVFRPDISPYFGLFYEKNGTNDGDYVFLTGEDSYLNFTKIVEWNGKTSLDWWITDKCNMINGTDGDSFHPLITKDEVLYVFPSDFCRSVYITFSDYESVQGLPAFRYKVPAEILANTSDNAGFCIPEGNCLGSGVLNVSICKNGAPIIMSFPHFYQADERFVSAIEGMHPNQEDHETFVDINPLTGIILKAAKRFQINIYVKKLDDFVETGDIRTMVFPVMYLNESVHIDKETASRLKSMINTTLIITNIPYIIMALGVFFGLVFTWLACKGQGSMDEGTADERAPLIRT

Specifications

Accession Number NP_005497.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 950
Molecular Weight (g/mol) 80.7kDa
Name SCARB2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MGRCCFYTAGTLSLLLLVTSVTLLVARVFQKAVDQSIEKKIVLRNGTEAFDSWEKPPLPVYTQFYFFNVTNPEEILRGETPRVEEVGPYTYRELRNKANIQFGDNGTTISAVSNKAYVFERDQSVGDPKIDLIRTLNIPVLTVIEWSQVHFLREIIEAMLKAYQQKLFVTHTVDELLWGYKDEILSLIHVFRPDISPYFGLFYEKNGTNDGDYVFLTGEDSYLNFTKIVEWNGKTSLDWWITDKCNMINGTDGDSFHPLITKDEVLYVFPSDFCRSVYITFSDYESVQGLPAFRYKVPAEILANTSDNAGFCIPEGNCLGSGVLNVSICKNGAPIIMSFPHFYQADERFVSAIEGMHPNQEDHETFVDINPLTGIILKAAKRFQINIYVKKLDDFVETGDIRTMVFPVMYLNESVHIDKETASRLKSMINTTLIITNIPYIIMALGVFFGLVFTWLACKGQGSMDEGTADERAPLIRT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AMRF/CD36L2/HLGP85/LIMPII/SR-BII
Common Name SCARB2
Gene Symbol SCARB2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.