Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD44v5 (Soluble) Human Instant ELISA™ Kit

Catalog No. 5018045
Encompass_Preferred
Change view
Click to view available options
Quantity:
128 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-180-45 128 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-180-45 Supplier Invitrogen™ Supplier No. BMS220INST

Includes

  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var(v5), HRP-Conjugate (human sCD44var(v5) monoclonal antibody) and Sample Diluent, lyophilized
  • Aluminum pouch(es) with a human sCD44var(v5) Standard curve (coloured)
  • Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
  • Sample Diluent (Use when an external predilution of the samples is needed)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Adhesive Films

Instant ELISA Kit

The Human Soluble CD44var (v5) ELISA quantitates Hu sCD44var (v5) in human serum, plasma, cell culture supernatants or other body fluids. The assay will exclusively recognize both natural and recombinant Hu sCD44var (v5). Principle of the method The Human sCD44var (v5) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). This protein participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis. [provided by RefSeq, Jul 2008]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16070
Assay Range 0.16-10.0 ng/mL
Assay Sensitivity 0.01 ng/mL
Conjugate HRP
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 960
Gene Symbol CDW44,CSPG8,ECMR-III,HCELL,HUTCH-I,IN,LHR,MC56,MDU2,MDU3,MIC4,Pgp1
Interassay CV 7.1%
Intraassay CV 7.2%
Kit Contents Pre-coated 96 well Microplate, Detection Antibody, Anti-IgG-HRP, Sample Diluent, lyophilized, Standard curve strips, Wash Buffer Concentrate, Substrate Solution, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye HRP
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL
Storage Requirements -20°C
Synonym VFF-8 Phagocytic glycoprotein I, Hermes antigen
Target Human
Test Time 20 min.
Total Assay Time 4 hr. 10 min.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.