Learn More
Invitrogen™ CD44v6 (Soluble) Human Instant ELISA™ Kit
Description
Includes
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v6), HRP-Conjugate (human sCD44var (v6) monoclonal antibody) and Sample Diluent, lyophilized
- Aluminum pouch(es) with a human sCD44var (v6) Standard curve (coloured)
- Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
- Sample Diluent
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (1M Phosphoric acid)
- Adhesive Films
The Human Soluble CD44var (v6) ELISA quantitates Hu sCD44var (v6) in human serum, plasma, cell culture supernatants, urine or other body fluids. The assay will exclusively recognize both natural and recombinant Hu sCD44var (v6). Principle of the method The Human sCD44var (v6) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P16070 |
| Assay Range | 0.16-10.0 ng/mL |
| Assay Sensitivity | 0.06 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA Kit |
| Sample Type | Plasma, Serum, Supernatant, Urine |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 960 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.