Learn More
Gibco™ Human SCF, Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P21583 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 4254 |
| Molecular Weight (g/mol) | 18.4 kDa |
| Name | Human SCF, Animal-Free |
| Quantity | 10 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.