Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human SCGF-alpha Recombinant Protein, PeproTech®

Catalog No. 10022A500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10022A500UG 500 μg
10022A100UG 100 μg
10022A10UG 10 μg
100-22A-1MG 1 mg
10022A250UG 250 μg
100-22A-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10022A500UG Supplier Gibco™ Supplier No. 10022A500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human SCGF-alpha is a 33.9 kDa polypeptide containing 305 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y240
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6320
Molecular Weight (g/mol) 33.9 kDa
Name Human SCGF-alpha
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
Common Name CLEC11A
Gene Symbol CLEC11A
Biological Activity Determined by its inhibitory effect on the proliferation of TF-1 cells previously grown in media containing GM-CSF. The ED50 for this effect was observed (at ≤ 2.5 ng/ml)
Conjugate Unconjugated
Recombinant Recombinant
Sequence GHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ AAAQARCTAR GGSLAQPADR QQMEALTRYL RAALAPYNWP VWLGVHDRRA EGLYLFENGQ RVSFFAWHRS PRPELGAQPS ASPHPLSPDQ PNGGTLENCV AQASDDGSWW DHDCQRRLYY VCEFPF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.