Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human SCGF-beta Recombinant Protein, PeproTech®

Catalog No. 10022B250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10022B250UG 250 μg
10022B100UG 100 μg
10022B10UG 10 μg
100-22B-1MG 1 mg
100-22B-2UG 2 μg
10022B500UG 500 μg
10022B50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 10022B250UG Supplier Gibco™ Supplier No. 10022B250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human SCGF-beta is a 25.0 kDa polypeptide containing 227 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y240
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6320
Molecular Weight (g/mol) 25 kDa
Name Human SCGF-beta
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
Common Name CLEC11A
Gene Symbol CLEC11A
Biological Activity Determined by the dose-dependent stimulation of the proliferation of TF-1 cells previously grown in media containing GM-CSF. The ED50 for this effect was observed (at ≤ 7 ng/ml)
Conjugate Unconjugated
Recombinant Recombinant
Sequence MGHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ PSASPHPLSP DQPNGGTLEN CVAQASDDGS WWDHDCQRRL YYVCEFPF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.