Learn More
Sino Biological Human SDHA PAb 100UL 104719-T42-100
Supplier:  Sino Biological 104719T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SDHA, and purified by antigen affinity chromatography.
Catalog No. 50-240-8217
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.