Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SEC13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7083302
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9055SP 25 μg
MAB9055100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9055SP Supplier R&D Systems Supplier No. MAB9055SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SEC13 Monoclonal specifically detects SEC13 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen SEC13
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1280A
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 0.3-25 ug/mL
Formulation Supplied as a solution in PBS
Gene Accession No. P55735
Gene Alias D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae)
Host Species Rabbit
Immunogen E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6396
Test Specificity Detects human SEC13 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.