Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SEMA6D Partial ORF (NP_705871.1, 852 a.a. - 940 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-420 10 μg
89-961-421 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-420 Supplier Abnova™ Supplier No. H00080031Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphorin domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. This gene encodes a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner. [provided by RefSeq]

Sequence: KLQNIDHPLTKSSSKRDHRRSVDSRNTLNDLLKHLNDPNSNPKAIMGDIQMAHQNLMLDPMGSMSEVPPKVPNREASLYSPPSTLPRNS

Specifications

Accession Number NP_705871.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80031
Molecular Weight (g/mol) 35.53kDa
Name SEMA6D (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen KLQNIDHPLTKSSSKRDHRRSVDSRNTLNDLLKHLNDPNSNPKAIMGDIQMAHQNLMLDPMGSMSEVPPKVPNREASLYSPPSTLPRNS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ11598/KIAA1479
Common Name SEMA6D
Gene Symbol SEMA6D
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.