Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Semaphorin 4G ELISA Kit

Catalog No. EH406RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH406RB 96 Tests
EH406RBX5 5 x 96 Tests
EH406RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH406RB Supplier Invitrogen™ Supplier No. EH406RB
Add to Cart
Add to Cart

ELISA

Human Semaphorin 4G quantitates human Semaphorin 4G in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin 4G.

Cell surface receptor for PLXNB2. May play a role in axon guidance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AI507908; AW554132; KIAA1619; KIAA1619FLJ20590; MGC102867; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4 G; SEMA4G; semaphorin 4 G; semaphorin-4 G
Assay Range 0.085 to 20 ng/mL
Assay Sensitivity 0.085 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 57715
Gene Symbol SEMA4G
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target SEMA4G
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.