Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 6A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1146SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1146SP 25 μg
MAB1146 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1146SP Supplier R&D Systems Supplier No. MAB1146SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Semaphorin 6A Monoclonal specifically detects Semaphorin 6A in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 6A
Applications Western Blot
Classification Monoclonal
Clone 169203
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H2E6
Gene Alias KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, Sema6A, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA
Gene Symbols SEMA6A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A Gly19-Thr649 Accession # Q9H2E6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57556
Test Specificity Detects human Semaphorin 6A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) Semaphorin 6A is observed and no cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 3B, or rmSemaphorin 3F is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.