Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SENP3 Partial ORF (NP_056485, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961432 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-432 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-432 Supplier Abnova™ Supplier No. H00026168Q01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates (Di Bacco et al., 2006 [PubMed 16738315]).[supplied by OMIM]

Sequence: EKAGQHSPLREEHVTCVQSILDEFLQTYGSLIPLSTDEVVEKLEDIFQQEFSTPSRKGLVLQLIQSYQRMPGNAMVRGFRVAYKRHVLTMDDLGTLYGQN

Specifications

Accession Number NP_056485
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26168
Molecular Weight (g/mol) 36.74kDa
Name SENP3 (Human) Recombinant Protein (Q01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen EKAGQHSPLREEHVTCVQSILDEFLQTYGSLIPLSTDEVVEKLEDIFQQEFSTPSRKGLVLQLIQSYQRMPGNAMVRGFRVAYKRHVLTMDDLGTLYGQN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp586K0919/DKFZp762A152/SMT3IP1/SSP3
Common Name SENP3
Gene Symbol SENP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.