Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SENP5 Full-length ORF (NP_689912.1, 1 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961436 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-436 10 μg
89-961-437 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-436 Supplier Abnova™ Supplier No. H00205564P01S

Used for AP, Array, ELISA, WB-Re

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for numerous biologic processes. SUMO-specific proteases, such as SENP5, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates (Di Bacco et al., 2006 [PubMed 16738315]).[supplied by OMIM]

Sequence: MKKQRKILWRKGIHLAFSEKWNTGFGGFKKFYFHQHLCILKAKLGRPVTWNRQLRHFQGRKKALQIQKTWIKDEHLCAKTKFNVATQNVSTLSSKVKRKDAKHFISSSKTLLRLQAEKLLSSAKNSDHEYCREKNLLKAVTDFPSNSALGQANGHRPRTDPQPSDFPMKFNGESQSPGESGTIVVTLNNHKRKGFCYGCCQGPEHHRNGGPLIPKKFQLNQHRRIKLSPLMMYEKLSMIRFRYRILRSQHFRTKSKVCKLRKAQRSWVQKVTGDHQETRRENGEGGSCSPFPSPEPKDPSCRHQPYFPDMDSSAVVKGTNSHVPDCHTKGSSFLGKELSLDEAFPDQQNGSATNAWDQSSCSSPKWECTELIHDIPLPEHRSNTMFISETEREIMTLGQENQTSSVSDDRVKLSVSGADTSVSSVDGPVSQKAVQNENSYQMEEDGSLKQSILSSELLDHPYCKSPLEAPLVCSGLKLENQVGGGKNSQKASPVDDEQLSVCLSGFLDEVMKKYGSLVPLSEKEVLGRLKDVFNEDFSNRKPFINREITNYRARHQKCNFRIFYNKHMLDMDDLATLDGQNWLNDQVINMYGELIMDAVPDKVHFFNSFFHRQLVTKGYNGVKRWTKKVDLFKKSLLLIPIHLEVHWSLITVTLSNRIISFYDSQGIHFKFCVENIRKYLLTEAREKNRPEFLQGWQTAVTKCIPQQKNDSDCGVFVLQYCKCLALEQPFQFSQEDMPRVRKRIYKELCECRLMD

Specifications

Accession Number NP_689912.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 205564
Molecular Weight (g/mol) 113.1kDa
Name Human SENP5 Full-length ORF (NP_689912.1, 1 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp564O1016, FLJ42398, MGC27076
Common Name SENP5
Gene Symbol SENP5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.