Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SEPSECS (NP_058651, 1 a.a. - 501 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89961456 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-456 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-456 Supplier Abnova™ Supplier No. P3415
Add to Cart
Add to Cart

Used for SDS-PAGE

The 21st amino acid, selenocysteine (sec), is distinct from other amino acids because it lacks its own tRNA synthetase and is the only amino acid synthesized on its cognate tRNA. Synthesis of sec begins with acylation of tRNA(sec) (TRSP; MIM 165060) by seryl-tRNA synthetase (SARS; MIM 607529) to give ser-tRNA(sec), which is subsequently phosphorylated by O-phosphoseryl-tRNA kinase (PSTK; MIM 611310) to give O-phosphoseryl-tRNA(sec). SEPSECS catalyzes the final step of sec synthesis by converting O-phosphoseryl-tRNA(sec) to selenocysteinyl-tRNA(sec) using selenophosphate as the selenium donor (Palioura et al., 2009 [PubMed 19608919]).[supplied by OMIM]

Sequence: MGSSHHHHHHSSGLVPRGSHMNRESFAAGERLVSPAYVRQGCEARRSHEHLIRLLLEKGKCPENGWDESTLELFLHELAIMDSNNFLGNCGVGEREGRVASALVARRHYRFIHGIGRSGDISAVQPKAAGSSLLNKITNSLVLDIIKLAGVHTVANCFVVPMATGMSLTLCFLTLRHKRPKAKYIIWPRIDQKSCFKSMITAGFEPVVIENVLEGDELRTDLKAVEAKVQELGPDCILCIHSTTSCFAPRVPDRLEELAVICANYDIPHIVNNAYGVQSSKCMHLIQQGARVGRIDAFVQSLDKNFMVPVGGAIIAGFNDSFIQEISKMYPGRASASPSLDVLITLLSLGSNGYKKLLKERKEMFSYLSNQIKKLSEAYNERLLHTPHNPISLAMTLKTLDEHRDKAVTQLGSMLFTRQVSGARVVPLGSMQTVSGYTFRGFMSHTNNYPCAYLNAASAIGMKMQDVDLFIKRLDRCLKAVRKERSKESDDNYDKTEDVDIEEMALKLDNVLLDTYQDASS

Specifications

Accession Number NP_058651
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 51091
Molecular Weight (g/mol) 57.9kDa
Name SEPSECS (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMNRESFAAGERLVSPAYVRQGCEARRSHEHLIRLLLEKGKCPENGWDESTLELFLHELAIMDSNNFLGNCGVGEREGRVASALVARRHYRFIHGIGRSGDISAVQPKAAGSSLLNKITNSLVLDIIKLAGVHTVANCFVVPMATGMSLTLCFLTLRHKRPKAKYIIWPRIDQKSCFKSMITAGFEPVVIENVLEGDELRTDLKAVEAKVQELGPDCILCIHSTTSCFAPRVPDRLEELAVICANYDIPHIVNNAYGVQSSKCMHLIQQGARVGRIDAFVQSLDKNFMVPVGGAIIAGFNDSFIQEISKMYPGRASASPSLDVLITLLSLGSNGYKKLLKERKEMFSYLSNQIKKLSEAYNERLLHTPHNPISLAMTLKTLDEHRDKAVTQLGSMLFTRQVSGARVVPLGSMQTVSGYTFRGFMSHTNNYPCAYLNAASAIGMKMQDVDLFIKRLDRCLKAVRKERSKESDDNYDKTEDVDIEEMALKLDNVLLDTYQDASS
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC161491/SLA/SLA/LP
Common Name SEPSECS
Gene Symbol SEPSECS
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.