Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SEPT4 Full-length ORF (NP_004565.1, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-472 25 ug
89-961-471 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-472 Supplier Abnova™ Supplier No. H00005414P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. The protein encoded by this gene is thought to be part of a complex involved in cytokinesis. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Sequence: MDRSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEAKTWASRPQVPEPRPQAPDLYDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVNSLFLTDLYRDRKLLGAEERIMQTVEITKHAVDIEEKGVRLRLTIVDTPGFGDAVNNTECWKPVAEYIDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPLDVEFMKALHQRVNIVPILAKADTLTPPEVDHKKRKIREEIEHFGIKIYQFPDCDSDEDEDFKLQDQALKESIPFAVIGSNTVVEARGRRVRGRLYPWGIVEVENPGHCDFVKLRTMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQEMLHKIQKQMKENY

Specifications

Accession Number NP_004565.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5414
Molecular Weight (g/mol) 81.5kDa
Name SEPT4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDRSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEAKTWASRPQVPEPRPQAPDLYDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVNSLFLTDLYRDRKLLGAEERIMQTVEITKHAVDIEEKGVRLRLTIVDTPGFGDAVNNTECWKPVAEYIDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPLDVEFMKALHQRVNIVPILAKADTLTPPEVDHKKRKIREEIEHFGIKIYQFPDCDSDEDEDFKLQDQALKESIPFAVIGSNTVVEARGRRVRGRLYPWGIVEVENPGHCDFVKLRTMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQEMLHKIQKQMKENY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ARTS/BRADEION/CE5B3/H5/MART/PNUTL2/SEP4/hCDCREL-2/hucep-7
Common Name SEPT4
Gene Symbol SEPT4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.