Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human Serpin E1/PAI-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Serpin E1/PAI-1 in cell culture supernates (50 ul), cell lysates (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul), ctad plasma (50 ul).
Specifications
Specifications
| Assay Range | 0.31 to 20 ng/mL |
| Assay Sensitivity | 0.142 ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Citrate, Heparin, CTAD) |
| Specificity | Natural and Recombinant Human Serpin E1 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 5054 |
| Quantity | 1 Pack (6 Plates) |
| Sample Volume | Cell Culture Supernates (50 uL),Cell Lysates (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL),Citrate Plasma (50 uL),CTAD Plasma (50 uL) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
